Please login to your online account to display your discounted pricing
GE Healthcare PhastGel™ Buffer Strips
Pre-prepared 3% agarose blocks containing buffer, for use in conjunction with PhastGel gradient and homogeneous gel media to generate a multiphasic buffer system during electrophoresis
$91.35 - $112.70
Specifications
For Use With (Equipment) PhastGel™ on PhastSystem automated flatbed electrophoresis units
View More Specs
Products
Catalog Number Mfr. No. For Use With (Application) Product Name Quantity Price Quantity    
45-000-476
ge healthcare
18166801
To hold Buffer strip Buffer strip holder 2 Pack Each for $112.70
45-000-090
ge healthcare
17051701
Used for industrial applications such as analytical chemistry and research PhastGel™ Buffer strips-native 20 Strips Pack of 20 Each for $91.35
45-000-089
ge healthcare
17051601
Used for industrial applications such as analytical chemistry and research PhastGel™ Buffer strips-SDS 20 Strips Pack of 20 Each for $91.35
Description & Specifications
Specifications
For Use With (Equipment) PhastGel™ on PhastSystem automated flatbed electrophoresis units
  • Ready made buffer strips eliminate the need to prepare electrophoresis buffers
  • Supplied as 10X pairs - for both cathode and anode
  • Choice of Native or Denaturing (SDS) buffer systems