Please login to your online account to display your discounted pricing
GE Healthcare Mono P Columns
Greatly concentrates samples and resolves pI differences of 0.02 pH units
$2,134.04 - $2,825.96
Specifications
Product Type Mono P ion exchange column
Material Borosilicate Glass
Support Type Monodisperse polystyrene/ divinyl benzene
Storage Requirements 4° to 30°C
View More Specs
Products
Catalog Number Mfr. No. Bed Volume For Use With (Application) Price Quantity    
45-500-112
ge healthcare
17517001
1mL Highest resolution separation Each for $2,134.04
45-000-306
ge healthcare
17-5171-01
4mL Rapid scouting of elution conditions (pH gradients) or for separations not requiring highest resolution Each for $2,825.96
Description & Specifications
Specifications
Product Type Mono P ion exchange column
Material Borosilicate Glass
Support Type Monodisperse polystyrene/ divinyl benzene
Storage Requirements 4° to 30°C
Mono P is based on the same matrix as Mono Q and Mono S. The 10µm MonoBeads particles are substituted with tertiary and quaternary amines and the small bead size contributes significantly to the high resolution that can be achieved.