Please login to your online account to display your discounted pricing

GE Healthcare MiniBeads™ Columns

MiniBeads™ prepacked columns are designed for high performance purification and characterization of biomolecules using ion exchange chromatography

$2,275.00 - $2,275.00

Specifications

Buffer All common Buffers except high-concentration Oxidizing and reactive agents, DMSO, DMF and Formic Acid
Quantity Each
Height (English) 1.96 in.
Height (Metric) 50mm
Particle Size 3μm
View More Specs
Products
Catalog Number Mfr. No. Flow Rate Ligand Type Material Max. Pressure Price Quantity    

45-000-307

 
ge healthcare
17517701
0.5 to 2mL/min. O-CH2-CHOH-CH2-O-CH2-CHOH-CH2-N+ (CH3) 3 Polyethere Ketone 180psi Each for $2,275.00
Description & Specifications

Specifications

Buffer All common Buffers except high-concentration Oxidizing and reactive agents, DMSO, DMF and Formic Acid
Quantity Each
Height (English) 1.96 in.
Height (Metric) 50mm
Particle Size 3μm
pH Range 3 to 11
Storage Requirements 4° to 30°C
Support Type Monodisperse Polystyrene/Divinyl Benzene particles

  • Small, mono-disperse beads for extremely high resolution purifications. non porous beads contribute to high resolution but give lower capacity than MonoBeads
  • Analytical and micro-preparative applications with choice of both anion and cation exchangers
  • High chemical and physical stability
  • Excellent reproducibility and durability

  • Tricorn high-performance columns (4.6/50 PE) prepacked with MiniBeads
  • PC columns designed for micropurification

Precision column holders may be required for use on high performance chromatography systems such as KTA design. For optimum results, it is important to check the compatibility of the column and the system specifications.