Please login to your online account to display your discounted pricing

GE Healthcare MBPTrap™ HP Prepacked Columns

Yield highly pure MBP-tagged recombinant proteins eluted in concentrated form and small volumes. GE Healthcare Life Sciences™ MBPTrap™ HP Prepacked Columns are prepacked with Dextrin Sepharose™ High Performance.

$346.00 - $1,426.00

Specifications

Bed Volume 1mL
Buffer Aqueous Buffer
Product Type Dextrin Sepharose HP prepacked column
For Use With (Application) Purification of MBP tagged Proteins
Height (Metric) 25mm
View More Specs
Products
Catalog Number Mfr. No. Flow Rate Packaging Quantity Price Quantity    

45-001-541

 
ge healthcare
28918779
<20mL/min. 1 x 5mL Each for $346.00

45-001-542

 
ge healthcare
28918780
<20mL/min. 5 x 5mL Pack of 5 Each for $1,426.00
Description & Specifications

Specifications

Bed Volume 1mL
Buffer Aqueous Buffer
Product Type Dextrin Sepharose HP prepacked column
For Use With (Application) Purification of MBP tagged Proteins
Height (Metric) 25mm
Ligand Type Dextrin
Material Polypropylene
Max. Pressure 3 bar
Particle Size 34μm
pH Range >7
Storage Requirements 2° to 8°C
Support Type 6% Highly Cross-linked Agarose

  • Prepacked with Dextrin Sepharose tagged with maltose binding protein (MBP).
  • Binding capacity is fusion protein dependent but can be as much as 16 mg/ml media
  • Maximum pressure: 3 bar
  • Compatible with commonly used buffers and easily regenerated with 0.5 M NaOH
  • 1 and 5mL polypropylene columns can be used either manually using a syringe or on an instrument

Media
  • Matrix: 6% cross-linked agarose
  • Average particle size: 34μm
  • pH stability working range: >7

1mL columns

  • Bed dimensions: 7 x 25mm
  • Flow velocity: <4mL/min.

5mL columns
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.