GE Healthcare Immobiline™ Sample Application Strips
Offer improved gel performance with faster and more stable gradients, fewer artifacts, and easier zymogram assays
$298.95 - $298.95
Specifications
Product Type SDS sample application strip
Quantity 5 Pk.
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Price Quantity  
45-001-796 GE Healthcare
18100274
26 SDS sample Pack of 5 Each for $298.95
Specifications
Description
  • For use with SDS and immobiline samples
  • Silicone applicator strip with sample holes to load samples onto Isoelectric focussing (IEF) Gels
Description & Specifications
Specifications
SDS sample application strip
5 Pk.