Please login to your online account to display your discounted pricing
GE Healthcare IMAC Sepharose™ High Performance Media
For optimizing purification of histidine-tagged proteins using Immobilized Metal Affinity Chromatography (IMAC) when Ni2+ is not the best choice of metal ion
$1,315.00 - $1,315.00
Products
Catalog Number Mfr. No. Price Quantity    
45-000-166
ge healthcare
17092007
Each for $1,315.00
Description & Specifications
  • 34μm beads of highly cross-linked 6% agarose, to which a chelating group has been covalently coupled
  • Charge with transition metal of choice for optimized selectivity
  • Small bead size for high performance purification
  • High binding capacity: ≥40mg histidine-tagged protein/mL medium when charged with Ni2+
  • Allows more sample to be applied; a more concentrated target protein is obtained more at a lower cost/mg and in less time
  • Stable in pH range 3-12 (long term), and in all commonly used buffers
Avoid use with chelating agents, e.g., EDTA, EGTA, or citrate.