Please login to your online account to display your discounted pricing
GE Healthcare illustra™ RNAspin 96 RNA Isolation Kit
For fast and efficient purification in a 96-well format
$1,439.64 - $1,439.64
Products
Catalog Number Mfr. No. Price Quantity    
45-001-757
ge healthcare
25050075
Pack of 4 Each for $1,439.64
Description & Specifications
  • 96 samples parallel-processed via either centrifugation or vacuum with high reproducibility in under 70 minutes
  • Can be used to isolate total RNA from cultured cells, tissue, bacteria or yeast
  • DNase I treatment removes genomic DNA resulting in high-quality total RNA product
  • Isolated RNA is of sufficient quality (A260/A280 = 1.8-2.2) and quantity (up to 100μg) for multiple downstream experiments: quantitative reverse transcription polymerase chain reaction (QRT-PCR), primer extension and RNase protection assays
  • Integrated wash plate eliminates risk of contamination
  • Accessory tissue plate allows tissue samples to be processed
  • Sample size: up to 30mg for animal tissue, up to 2 x 106 cells for cell culture (vaccum); 1x 107 cells for cell culture (centrifuge); Elution volume: 50μ-130μL