Please login to your online account to display your discounted pricing
GE Healthcare illustra™ CyScribe GFX™ Purification Kit
Specifically developed for the rapid purification of CyDye labeled cDNA probes
$297.90 - $297.90
Specifications
For Use With (Application) Rapid purification of Cydye-labeled cDNA probes
For Use With (Equipment) CyScribe™ microarray labeling kits
Product Name Kit
View More Specs
Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    
45-001-423
ge healthcare biosciences
27960602
25mL Capture Buffer type 1, 30mL Wash Buffer type 3, 16mL Elution Buffer type 7, 50 Illustra™ CyScribe™ GFX™ MicroSpin™ columns, 50 collection tubes 50 Preps Each for $297.90
Description & Specifications
Specifications
For Use With (Application) Rapid purification of Cydye-labeled cDNA probes
For Use With (Equipment) CyScribe™ microarray labeling kits
Product Name Kit
  • Provides efficient removal of unincorporated CyDye label and primers from labeling reactions with superior yields of labeled cDNA probe
  • Excellent for the purification of cDNA labeled by either direct incorporation or by post-labeling using CyScribe labeling kits
  • Optimized purification reagents available in both 25- and 50-column format; 50-column included with CyScribe labeling kits