Please login to your online account to display your discounted pricing
Hoefer™ PR 648 Slot Blot Blotting Manifold
Provides a simple, convenient method for screening recombinant clones, DNA, RNA and proteins
$1,695.38 - $1,695.38
Specifications
Material Acrylic
No. of Slots 48.0
Temperature (English) Maximum 113°F
Temperature (Metric) Maximum 45°C
Volume (Metric) Sample 1mL
View More Specs
Includes:  Top block, Membrane support block with O-ring, Bottom block with connector, QuickFit connector with O-ring, Six clamping screws, Membrane cutting template
Products
Catalog Number Mfr. No. Includes Price Quantity    
45-002-241
ge healthcare
80609558
Top block, membrane support block with O-ring, bottom block with connector, QuickFit™ connector with O-ring, 6 clamping screws, membrane cutting template Each for $1,695.38
Description & Specifications
Specifications
Material Acrylic
No. of Slots 48.0
Temperature (English) Maximum 113°F
Temperature (Metric) Maximum 45°C
Volume (Metric) Sample 1mL
Forty-eight slots in a 4 × 12 grid empty into a single lower chamber which has a fitting for attaching a vacuum pump or aspirator to draw samples through the slot. Each slot is 6mm long, 0.80mm wide, and exposes 4.8mm2 of membrane.
  • Raised lip around each slot creates an effective seal and prevents cross-contamination between wells
  • Alphanumeric grid facilitates accurate sample application
  • Alignment pins ensure quick and easy assembly
  • Membrane blotted in the Slot Blot, or autoradiograms made from the membranes, are easily quantified by densitometry
  • Sturdy acrylic construction