Hoefer™ Combs for HE 33 Mini Submarine Unit
Accessory parts for HE 33 Mini Submarine
Manufacturer:  Hoefer™ HE31A161.5
 View more versions of this product
Catalog No. 36-100-3918
Description
Description
Specifications
Specifications
0.19 in.
HE33 Mini Submarine Unit
16
0.15 cm
0.1 in.
0.5 cm
Backless Comb
0.05 in.
20.5 μL
0.26 cm