Please login to your online account to display your discounted pricing

GE Healthcare HiTrap™ Protein G HP Prepacked Columns Promotion

For a limited time only, receive a 10% discount on your next HiTrap order

$693.28 - $2,831.17

Specifications

For Use With (Application) Fast, easy separations of IgG from different species and subclasses.
Storage Buffer 20% Ethanol
Storage Requirements 4° to 30°C
View More Specs
Products
Catalog Number Mfr. No. Binding Capacity Diameter (English) Inner Column Flow Rate Volume (Metric) Bed Price Quantity    

45-000-053PM

 
ge healthcare
29133586
25mg Human IgG/column 0.27 in. <4mL/min. Bed Dimensions: 7 x 25mm; 5 x 1mL Pack of 5 Each for $693.28

45000056PM

 
ge healthcare
17040503PM
125mg Human IgG/column 0.62 in. <20mL/min. Bed Dimensions: 16 x 25mm; 5 x 5mL Pack for $2,831.17
Description & Specifications

Specifications

For Use With (Application) Fast, easy separations of IgG from different species and subclasses.
Storage Buffer 20% Ethanol
Storage Requirements 4° to 30°C

 

  • Prepacked with Protein G Sepharose High Performance
  • Binding capacity: 25mg human IgG/column
  • Maximum pressure: 3 bar
  • Column material: polypropylene
  • For use with syringe, peristalic pump, or chromatography system
  • 1 and 5mL columns
Media
  • Ligand: Recombinant protein G lacking albumin-binding region
  • Matrix: 6% cross-linked agarose
  • Average particle size: 34μm
  • pH stability working range: 3 to 9

 

 

  • Cannot be combined with any other discounts
  • Offer expires June 30, 2016

 

1mL columns

  • Bed dimensions: 7 x 25mm
  • Flow velocity: <4mL/min.
5mL columns
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.