Please login to your online account to display your discounted pricing

GE Healthcare HiTrap™ Desalting and HisTrap™ Pre-packed Columns: Promo Offer

Special offer is available 1/1/2016 through 3/31/2016

$66.25 - $66.25

Specifications

Storage Requirements 4 to 30°C
View More Specs
Products
Catalog Number Mfr. No. Description Product Type Quantity Price Quantity    

45000511PM

 
ge healthcare™
29048684PM
Packaging Quantity: 1 x 5mL HiTrap Desalting Columns Each Each for $66.25
Description & Specifications

Specifications

Storage Requirements 4 to 30°C

10% Savings on HiTrap™ Desalting and HisTrap™ Pre-packed Columns!

HiTrap Desalting is a ready-to-use 5mL column, prepacked with Sephadex™ G-25 Superfine, for fast and convenient desalting and buffer exchange. Handles sample volumes of 1.5mL, or larger volumes by connecting up to five columns in series. Replaces dialysis as the method of choice for desalting.

HiTrap Desalting Columns: 45-002-908PM, 45-000-511PM
HisTrap Excel Columns: 45-000-505PM


  • For rapid convenient desalting and buffer exchange
  • Prepacked with Sephadex™ G-25 Superfine media
  • Column material: polypropylene
  • Bed volume: 5mL
  • Bed dimensions: 16 x 25mm
  • Sample volume: 1.5mL
  • Flow velocity: <15mL/min.
  • Maximum pressure: 3 bar


Media
  • Matrix: cross-linked dextran
  • Particle size: >20μm
  • pH stability working range: 2 to 13
Storage: 4° to 30°C in 20% ethanol.