missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva HiTrap™ AVB Sepharose™ Columns
Enables purification of adeno associated viruses (AAV) in one step, with high purity and excellent yields. Cytiva HiTrap™ AVB Sepharose™ Columns are pre-packed with affinity chromatography medium (resin) for simple operation with a syringe, pump or chromatographic system.
$1951.24 - $2018.57
Specifications
For Use With (Application) | One Step Purification of Adeno Associated Viruses |
---|---|
Stability | Stable to all commonly used aqueous buffers |
Storage Requirements | Store prepacked column at 4°C to 30°C in 20% Ethanol and Media at 4°C to 8°C in 20% Ethanol. |
Binding Capacity | >10â¿12 genecopies/mL Chromatography Mediμm |
Product Type | HiTrap AVB Sepharose HP Column |
Description
AVB Sepharose High Performance is an affinity resin designed for the purification of adeno associated virus (AAV). It enables purification of adeno associated viruses in one step with high purity and excellent yields.- One step purification of adeno associated viruses
- Affinity resin with high selectivity and low background binding
- Large surface area to give enhanced capacity
- Prepacked HiTrap columns for simple operation with a syringe, pump, or chromatographic system
- It is expected to have affinity for most of the other adeno associated virus subclasses
- The ligand is a 14 kD fragment from a single chain antibody, expressed in yeast
Specifications
One Step Purification of Adeno Associated Viruses | |
Store prepacked column at 4°C to 30°C in 20% Ethanol and Media at 4°C to 8°C in 20% Ethanol. | |
HiTrap AVB Sepharose HP Column | |
24 to 44 μm | |
Polypropylene | |
Cross-linked agarose | |
14 kD recombinant protein |
Stable to all commonly used aqueous buffers | |
>10â¿12 genecopies/mL Chromatography Mediμm | |
<150 cm/hr | |
25 mm | |
70 psi (5 bar) | |
3 to 10 |