Please login to your online account to display your discounted pricing

GE Healthcare HiPrep™ IEX FF 16/10 Columns

Provide fast, reliable and reproducible preparative ion exchange separations

$788.00 - $788.00

Specifications

Bed Volume 20mL
Buffer 1M NaOH, 8M Urea, 8M Guanidine Hydrochloride, 70% Ethanol
Diameter (English) Inner 0.62 in.
Diameter (Metric) Inner 16mm
Quantity 1 x 20mL
View More Specs
Products
Catalog Number Mfr. No. Ligand Type Max. Pressure Product Type pH Range Price Quantity    

45-002-066

 
ge healthcare
28936541
Diethylaminoethyl 5 bar HiPrep DEAE Sepharose FF column 2 to 12 Each for $788.00
Description & Specifications

Specifications

Bed Volume 20mL
Buffer 1M NaOH, 8M Urea, 8M Guanidine Hydrochloride, 70% Ethanol
Diameter (English) Inner 0.62 in.
Diameter (Metric) Inner 16mm
Quantity 1 x 20mL
Flow Rate >300cm/hr.
For Use With (Application) Preparative separations of Proteins and other biomolecules using ion exchange (IEX) chromato graphy
Height (English) 3.93 in.
Height (Metric) 100mm
Material Polypropylene
Particle Size 45 to 165μm
Storage Requirements 4° to 30°C
Support Type 6% Cross-linked Agarose

DEAE and CM Sepharose™ 6 Fast Flow are prepacked in 20mL HiPrep columns. Ideal for rapid enrichments during initial capture of proteins from starting material.

Features:

  • Combine good flowrates and high binding capacity
  • Simple-to-operate
  • Compatible with single-pump configurations and other chromatography systems (e.g., AKTA™)

Specifications (H2O at room temp.)

  • Dimensions: 1.6 x 10cm
  • Recommended flowrate: 2-10mL/min. (60-300cm/h)
  • Maximum flowrate: 10mL/min. (300cm/hr.)
  • Max. pressure over packed bed: 1.5bar (22psi, 0.15MPa)
  • Column hardware pressure limit: 5bar (73psi, 0.5MPa)
  • Storage: 20% ethanol (DEAE,CM), 0.2M sodium acetate in 20% ethanol (SP)
  • Storage temperature: 4° to 30°C