missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GE Healthcare High Purity Solution ddNTPs

Use in any application that requires ddNTPs of high purity. GE Healthcare Life Sciences™ High Purity Solution ddNTPs are available either as individual products or in a convenient set.

$274.80 - $287.00

Specifications

Color Colorless
Format Liquid
For Use With (Application) Dideoxy sequencing, genotyping reactions
Purity ≥98%
Concentration 100mM
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
45-001-340 GE Healthcare
27205101
ddATP Each for $287.00
45-001-344 GE Healthcare
27207101
ddGTP Each for $274.80
45-001-342 GE Healthcare
27206101
ddCTP Each for $279.00
Specifications

Description

  • Each lot of ddNTPs is functionally tested to meet or exceed criteria for high-quality sequencing with Thermo Sequenase™ DNA polymerase
  • High purity: >98% triphosphate

Dideoxy sequencing, Genotyping reactions

Description & Specifications

Specifications

Colorless
Dideoxy sequencing, genotyping reactions
100mM
Liquid
≥98%
4μmol (40μL)