Please login to your online account to display your discounted pricing
GE Healthcare GeneQuant™ 100 and 1300 Spectrophotometers Accessories
Manufacturer: ge healthcare  80210368
 View more versions of this product
Catalog No. BC80-2103-68
  • / Each
Description & Specifications
Specifications
Product Type Ultra-microcell Sample cell
Material Quartz
Volume (Metric) 5 to 7μL
For Use With (Equipment) GeneQuant 100 and 1300 spectrophotometers