Please login to your online account to display your discounted pricing

GE Healthcare GammaBind™ G Sepharose™ Medium

Use recombinant GammaBind™ for laboratory scale antibody purification. GE Healthcare Life Sciences™ GammaBind™ G Sepharose™ Medium binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse

$814.00 - $814.00

Specifications

Binding Capacity >18mg Human IgG/mL, 6mg Mouse IgG/mL
Product Type Sepharose Medium
For Use With (Application) For protein purification by affinity coupling
Ligand Concentration ≈3mg GammaBind G, Type 2/mL drained gel
Particle Size 45μm-165μm
View More Specs
Products
Catalog Number Mfr. No. Volume (Metric) Price Quantity    

45-000-157

 
ge healthcare
17088501
5mL Each for $814.00
Description & Specifications

Specifications

Binding Capacity >18mg Human IgG/mL, 6mg Mouse IgG/mL
Product Type Sepharose Medium
For Use With (Application) For protein purification by affinity coupling
Ligand Concentration ≈3mg GammaBind G, Type 2/mL drained gel
Particle Size 45μm-165μm
Storage Buffer PBS pH 7, 20% Ethanol

Binds to all IgG subclasses from human, mouse, and rat and total IgG from guinea pig, rabbit, goat, cow, sheep, and horse. Engineered for antibody purification at laboratory scale.

  • Will not cross-react with other serum proteins such as IgM, IgE, IgA or transferrin
  • Ligand: GammaBind G type 2 lacking albumin-binding region
  • Ligand coupling method: cyanogen bromide activation
  • Ligand density: ≈ 3mg GammaBind G, type 2/mL medium
  • Number of IgG binding sites per ligand: 2
  • Matrix: Agarose, 4%
  • Average particle size: 90μm
  • Binding capacity: >18 mg human IgG/ml, 6 mg mouse IgG/ml
  • pH stability: 3-9 (working), 2-9 (short term)