missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva (Formerly GE Healthcare Life Sciences) Ficoll PM 70 Centrifugation Media
Behave as ideal neutral spheres. Cytiva™ Ficoll PM70 Centrifugation Media is considered the molecules of choice for studying pore size distribution and the permeability of membranes.
$354.00 - $354.00
Specifications
Product Type | Centrifugation Media |
---|---|
For Use With (Application) | Specifically intended for use in perfusion media for laboratory studies on dextran-sensitive animals. |
Format | Powder |
Storage Requirements | 4–30°C |
Endotoxin Level | 0.12 EU/mg |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
45-002-020
|
Cytiva (Formerly GE Healthcare Life Sciences)
17031010 |
100 g |
Each for $354.00
|
|
Description
- High molecular weight (70,000) sucrose-polymer formed by copolymerization of sucrose with epichlorohydrin
- Highly branched structure and high content of hydroxyl groups mean good solubility in aqueous media with concentrations up to 50% w/v easily attained
- Displays minimal toxicity, plus favorable viscosity and osmotic properties
- Low endotoxin levels (<0.12 EU/mg)
For use in perfusion media for laboratory studies on dextran-sensitive animals
Order Info
Supplied as spray-dried powders
Specifications
Centrifugation Media | |
Powder | |
0.12 EU/mg | |
≈10mL/g |
Specifically intended for use in perfusion media for laboratory studies on dextran-sensitive animals. | |
4–30°C | |
White |