Please login to your online account to display your discounted pricing

GE Healthcare ExcelGel Precast Gels for Second Dimension Separations

Prepare multiple 2-D maps on a single gel using three 7cm or two 11cm IPG strips side-by-side

$289.00 - $289.00

Specifications

Color Colorless
Thickness (English) 0.019 in.
For Use With (Application) SDS-PAGE
Thickness (Metric) 0.05cm
View More Specs
Products
Catalog Number Mfr. No. Buffer Separation Range Stacking Zone Storage Requirements Price Quantity    

45-000-214

 
ge healthcare
17123601
Tris-Acetate, pH 6.6 12 to 116Mr 40mm, 5% T/3% C 4° to 30°C Pack of 3 Each for $289.00
Description & Specifications

Specifications

Color Colorless
Thickness (English) 0.019 in.
For Use With (Application) SDS-PAGE
Thickness (Metric) 0.05cm

  • Precast SDS-polyacrylamide gels available in 12.5% homogeneous and 12-14% gradient form
  • Perform second-dimension separations using precast gels together with 7, 11, 18, or 24 cm IPG strips (not included)
  • Provided fully hydrated and ready to use
  • Configure Multiphor II with MultiTemp III Thermostatic Circulator and the programmable EPS 1001 power supply for reproducible running conditions

Order IPG strips separately.