Please login to your online account to display your discounted pricing

GE Healthcare ExcelGel Precast Gels for SDS-PAGE

Save time with ready-to-use, mechanically stable gels cast on plastic supports. GE Healthcare Life Sciences™ ExcelGel Precast Gels for SDS-PAGE are available as homogeneous or gradient gels.

$634.00 - $970.00

Specifications

Color Colorless
Length (English) Gel 4.3 in.
Material Polyacrylamide
Thickness (English) 0.019 in.
For Use With (Application) Analytical chemistry, Research Horizontal electrophoresis of SDS denatured proteins
View More Specs
Products
Catalog Number Mfr. No. Format Separation Range Price Quantity    

45-001-633

 
ge healthcare
80126101
Precast Gel 15 kd Pack of 6 Each for $970.00

45-001-630

 
ge healthcare
80125553
Precast Gel 200 kd Each for $634.00
Description & Specifications

Specifications

Color Colorless
Length (English) Gel 4.3 in.
Material Polyacrylamide
Thickness (English) 0.019 in.
For Use With (Application) Analytical chemistry, Research Horizontal electrophoresis of SDS denatured proteins
Length (Metric) Gel 11cm
Thickness (Metric) 0.05cm

Three homogeneous and one gradient polyacrylamide gel for horizontal electrophoresis of SDS denatured proteins; also for separation of nucleic acids in the base pair range 50-1500

  • Homogeneous 7.5, 12.5, and 15% gels have 25 preformed sample wells for sample volumes from up to 17μL for quick, easy sample application
  • Gradient gel uses sample application strips (26 samples up to 40μL, 52 samples up to 20μL), paper pieces, or sample droplets; recommended for use with Immobiline DryStrip gels
  • ExcelGel SDS Homogeneous 15% is also designed for separation of peptides