Please login to your online account to display your discounted pricing

GE Healthcare Dithiothreitol (DTT)

Include reducing agents in the sample solution to break any disulfide bonds present and to maintain all proteins in their fully reduced state. GE Healthcare Life Sciences™ Dithiothreitol (DTT) is one of the most commonly used reductants.

$52.19 - $123.67

Specifications

For Use With (Application) IEF, SDS-PAGE
pH 0 to 5.1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

45-000-232

 
ge healthcare
17131801
1000mg Each for $52.19

45-000-233

 
ge healthcare
17131802
2000mg Each for $123.67
Description & Specifications

Specifications

For Use With (Application) IEF, SDS-PAGE
pH 0 to 5.1

  • Use for IEF, SDS-PAGE
  • White
  • Solid
  • Easily soluble in cold water and hot water
  • Threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent, DTT