Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon siRNA Ancillary reagents
$110.96 - $110.96
Products
Catalog Number Mfr. No. For Use With (Application) Price Quantity    
B002000
ge healthcare dharmacon
B-002000-UB-100
Resuspension and long-term storage of any short, double-stranded or single-stranded synthetic RNA molecule, provides long-term storage of siRNAs, RNAs, microRNA reagents Each for $110.96
Description & Specifications
Ancillary reagents for experimental workflow include:
siRNA Controls
A wide variety of recommended control products for every siRNA experiment
5X siRNA Buffer:
Appropriate for resuspension and long-term storage of any short, double-stranded or single-stranded synthetic RNA molecule, including modified and unmodified siRNAs, microRNA Mimics and microRNA Hairpin Inhibitors. Composition: 300mM KCl, 30mM HEPES-pH 7.5, 1.0mM MgCl2
RNase-free water:
Molecular Grade RNase-free water for dilution of 5x buffer
Accell Delivery Media:
Accell siRNA delivery media is an enriched, serum-free media specially formulated and optimized for Accell siRNA delivery.
DharmaFECT Cell Culture Reagent:
DharmaFECT Cell Culture Reagent (DCCR) is used to dilute DharmaFECT siRNA transfection reagent prior to use in Dharmacon Reverse Transfection format (RTF) siRNA library plates