Please login to your online account to display your discounted pricing

GE Healthcare Dharmacon siGLO™ Red Transfection Indicator

Chemically modified to localize to the nucleus and significantly extend fluorescence as a qualitative indicator of delivery and should not be used for quantitative determination of siRNA uptake.

Manufacturer: ge healthcare dharmacon  D0016300220

 View all options for this product

Catalog No. D0016300220


Description & Specifications


Fluorophore Cy3 Thodamine or Polyethylene
For Use With (Application) Determination of optimal siRNA transfection conditions in mammalian cells
Quantity 20nm

siGLO Red (DY-547) has a stronger signal at lower concentration, and is suited for electroporation or nucleofection.
Filter: CyTM3, Rhodamine, or PE, Absorbance: 557, Emission Max: 570 nm

  • Specially modified, fluorescent RNA duplex for simplified transfection assessment - without performing a functional assay
  • Fluorescent signal localizes to the nucleus as an unmistakable signal of uptake
  • Use as an indicator of transfection success in any RNAi experiment