Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon siGLO™ Red Transfection Indicator
Chemically modified to localize to the nucleus and significantly extend fluorescence as a qualitative indicator of delivery and should not be used for quantitative determination of siRNA uptake.
$239.49 - $1,047.19
Specifications
For Use With (Application) Determination of optimal siRNA transfection conditions in mammalian cells
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    
D0016300205
ge healthcare dharmacon
D-001630-02-05
5nm Each for $239.49
D0016300220
ge healthcare dharmacon
D0016300220
20nm Each for $592.03
D0016300250
ge healthcare dharmacon
D0016300250
50nm Each for $1,047.19
Description & Specifications
Specifications
For Use With (Application) Determination of optimal siRNA transfection conditions in mammalian cells
siGLO Red (DY-547) has a stronger signal at lower concentration, and is suited for electroporation or nucleofection.
Filter: CyTM3, Rhodamine, or PE, Absorbance: 557, Emission Max: 570 nm
  • Specially modified, fluorescent RNA duplex for simplified transfection assessment - without performing a functional assay
  • Fluorescent signal localizes to the nucleus as an unmistakable signal of uptake
  • Use as an indicator of transfection success in any RNAi experiment