Please login to your online account to display your discounted pricing

GE Healthcare Dharmacon siGLO™ Green Transfection Indicator

Chemically modified to localize to the nucleus and significantly extend fluorescence as a qualitative indicator of delivery and should not be used for quantitative determination of siRNA uptake.

Manufacturer: ge healthcare dharmacon  D0016300120

 View all options for this product

Catalog No. D0016300120


Description & Specifications


Fluorophore FITC
For Use With (Application) Determination of optimal siRNA transfection conditions in mammalian cells
Quantity 20nm

siGLO Green (FAM) exhibits nearly 100% signal localization to the nucleus, and represents an ideal choice for lipid-mediated transfection, followed by microscopy or FACS.
Filter: FITC, Absorbance: 494 nm, Emission Max: 520 nm

  • Specially modified, fluorescent RNA duplex for simplified transfection assessment - without performing a functional assay
  • Fluorescent signal localizes to the nucleus as an unmistakable signal of uptake
  • Use as an indicator of transfection success in any RNAi experiment