Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon siGLO™ Green Transfection Indicator
Chemically modified to localize to the nucleus and significantly extend fluorescence as a qualitative indicator of delivery and should not be used for quantitative determination of siRNA uptake.
$239.49 - $592.03
Specifications
For Use With (Application) Determination of optimal siRNA transfection conditions in mammalian cells
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    
D0016300105
ge healthcare dharmacon
D0016300105
5nm Each for $239.49
D0016300120
ge healthcare dharmacon
D0016300120
20nm Each for $592.03
Description & Specifications
Specifications
For Use With (Application) Determination of optimal siRNA transfection conditions in mammalian cells
siGLO Green (FAM) exhibits nearly 100% signal localization to the nucleus, and represents an ideal choice for lipid-mediated transfection, followed by microscopy or FACS.
Filter: FITC, Absorbance: 494 nm, Emission Max: 520 nm
  • Specially modified, fluorescent RNA duplex for simplified transfection assessment - without performing a functional assay
  • Fluorescent signal localizes to the nucleus as an unmistakable signal of uptake
  • Use as an indicator of transfection success in any RNAi experiment