Please login to your online account to display your discounted pricing

GE Healthcare Dharmacon siGLO™ Cyclophilin B Control siRNA

Silences the Cyclophilin B gene and is labeled with DY-547

Manufacturer: ge healthcare dharmacon  D0016100105

 View all options for this product

Catalog No. D0016100105

 Please call Customer Service at 1-800-234-7437 or send an email to for assistance.

Description & Specifications


Accession Number NM_000942 (Human) , NM_011149 (Mouse) and NM_022536 (Rat)
Fluorophore Cy3 Thodamine or Polyethylene
For Use With (Application) Validated positive control, guaranteed to silence cyclophilin B in Human, Mouse or Rat cells
Product Type siGLO Cyclophilin B siRNA
Quantity 5nmol

Stable, fluorescent positive control suitable for RNAi experiments in human, mouse/rat cells

  • Targets accession numbers NM_000942 (Human), NM_011149 (Mouse)/NM_022536 (Rat)
  • Labeled with DY-547
  • Absorbance/Emission Max is 557/570nm
  • A Cy3, Rhodamine or PE filter can be used
  • Significantly extended intensity of the fluorescence signal (7 days or longer)
  • Positive correlation of fluorescence with transfection efficiency and target gene silencing