Please login to your online account to display your discounted pricing

GE Healthcare Dharmacon siGLO™ Cyclophilin B Control siRNA

Silences the Cyclophilin B gene and is labeled with DY-547

Specifications

Accession Number NM_000942 (Human) , NM_011149 (Mouse) and NM_022536 (Rat)
Product Type siGLO Cyclophilin B siRNA
For Use With (Application) Validated positive control, guaranteed to silence cyclophilin B in Human, Mouse or Rat cells
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

D0016100105

 
ge healthcare dharmacon
D0016100105
5nmol CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Accession Number NM_000942 (Human) , NM_011149 (Mouse) and NM_022536 (Rat)
Product Type siGLO Cyclophilin B siRNA
For Use With (Application) Validated positive control, guaranteed to silence cyclophilin B in Human, Mouse or Rat cells

Stable, fluorescent positive control suitable for RNAi experiments in human, mouse/rat cells

  • Targets accession numbers NM_000942 (Human), NM_011149 (Mouse)/NM_022536 (Rat)
  • Labeled with DY-547
  • Absorbance/Emission Max is 557/570nm
  • A Cy3, Rhodamine or PE filter can be used
  • Significantly extended intensity of the fluorescence signal (7 days or longer)
  • Positive correlation of fluorescence with transfection efficiency and target gene silencing