Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon siGENOME™ Non-Targeting Control siRNAs
Negative control siRNAs designed to target no known genes in human, mouse or rat
$174.04 - $481.94
Specifications
For Use With (Application) Designed to have comparable GC content to that of functional siRNA, Used in siRNA-mediated RNAi experiments as controls for nonsequence specific effects and determination of baseline cellular responses in RNAi experiments
View More Specs
Products
Catalog Number Mfr. No. Product Type Quantity Price Quantity    
D0012100205
ge healthcare dharmacon
D0012100205
siCONTROL Nontargeting siRNA #2 5nmol Each for $174.04
D0012100220
ge healthcare dharmacon
D0012100220
siCONTROL Nontargeting siRNA #2 20nmol Each for $481.94
Description & Specifications
Specifications
For Use With (Application) Designed to have comparable GC content to that of functional siRNA, Used in siRNA-mediated RNAi experiments as controls for nonsequence specific effects and determination of baseline cellular responses in RNAi experiments
siGENOME Non-Targeting Control siRNAs are highly functional, chemically synthesized, and mostly unmodified negative control siRNA duplexes for experiments in human, mouse, and rat cells. siGENOME Non-Targeting Controls were designed to have a minimum of 4 mismatches to all human, mouse and rat genes, and confirmed to have minimal targeting by genomewide microarray analysis. Changes in mRNA or protein levels in cells treated with these controls reflect a baseline cellular response that can be compared to the levels in cells treated with target-specific siRNA.
  • BLAST analysis confirms at least 4 mismatches with all known human, mouse, and rat genes
  • Screened by microarray analysis
  • siGENOME Non-targeting siRNA - 1 reduces EGFR mRNA (NM_005228) in an assay- and cell-specific manner by ∽50%. Originally reported by customers, this finding was not observed by initial microarray validation, but has been subsequently verified by internal scientists. Recommended only if you have successfully used this control in past experiments
  • siGENOME Non-Targeting siRNA - 2 and 4 target firefly luciferase mRNA (U47296)
  • siRNAs can be used either as non-targeting siRNA in human, mouse/rat cell lines or as positive silencing control in systems using firefly luciferase as reporter gene (pGL3 cloning vector, Promega)
  • siGENOME Non-Targeting siRNA - 5 carries the ON-TARGET modification pattern to prevent sense strand uptake
Recommended for determination of baseline cellular responses in RNAi experiments