Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon ON-TARGETplus™ Non-Targeting siRNAs
Manufacturer: ge healthcare dharmacon  D-001810-01-05
 View more versions of this product
Catalog No. D0018100105
  • / Each
Description & Specifications
Specifications
For Use With (Application) Ideal negative control for RNAi experiments in human, mouse and rat cells, chemically matched for use with ON-TARGETplus siRNA reagents, determination of baseline cellular responses in RNAi experiments
Product Type ON-TARGETplus Non-Targeting SiRNA#1
Quantity 5nm
ON-TARGETplus designed, modified, and microarray-confirmed to have minimal targeting of known genes in human, mouse and rat cells
  • Ideal negative control for RNAi experiments in human, mouse, or rat cells
  • Chemically matched for use with ON-TARGETplus siRNA reagents