Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon ON-TARGETplus™ Non-Targeting siRNAs
$212.71 - $542.94
Specifications
For Use With (Application) Ideal negative control for RNAi experiments in human, mouse and rat cells, chemically matched for use with ON-TARGETplus siRNA reagents, determination of baseline cellular responses in RNAi experiments
View More Specs
Products
Catalog Number Mfr. No. Product Type Quantity Price Quantity    
D0018100105
ge healthcare dharmacon
D-001810-01-05
ON-TARGETplus Non-Targeting SiRNA#1 5nm Each for $212.71
D0018100120
ge healthcare dharmacon
D-001810-01-20
ON-TARGETplus Non-Targeting SiRNA#1 20nm Each for $542.94
Description & Specifications
Specifications
For Use With (Application) Ideal negative control for RNAi experiments in human, mouse and rat cells, chemically matched for use with ON-TARGETplus siRNA reagents, determination of baseline cellular responses in RNAi experiments
ON-TARGETplus designed, modified, and microarray-confirmed to have minimal targeting of known genes in human, mouse and rat cells
  • Ideal negative control for RNAi experiments in human, mouse, or rat cells
  • Chemically matched for use with ON-TARGETplus siRNA reagents