Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon ON-TARGETplus™ Non-Targeting Pool
Manufacturer: ge healthcare dharmacon  D-001810-10-05
 View more versions of this product
Catalog No. D0018101005
  • / Each
Description & Specifications
Specifications
For Use With (Application) Determination of baseline cellular responses in RNAi experiments, screened by genome-wide microarray analysis for minimal off-targets
Quantity 5nm
ON-TARGETplus designed, modified, and microarray-confirmed to have minimal targeting of known genes in human, mouse and rat cells
  • Pool of four non-targeting siRNAs
  • Ideal negative control for RNAi experiments using human, mouse or rat SMARTpool reagents.