Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon ON-TARGETplus™ Non-Targeting Pool
$254.36 - $595.00
Specifications
For Use With (Application) Determination of baseline cellular responses in RNAi experiments, screened by genome-wide microarray analysis for minimal off-targets
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    
D0018101005
ge healthcare dharmacon
D-001810-10-05
5nm Each for $254.36
D0018101020
ge healthcare dharmacon
D-001810-10-20
20nm Each for $595.00
Description & Specifications
Specifications
For Use With (Application) Determination of baseline cellular responses in RNAi experiments, screened by genome-wide microarray analysis for minimal off-targets
ON-TARGETplus designed, modified, and microarray-confirmed to have minimal targeting of known genes in human, mouse and rat cells
  • Pool of four non-targeting siRNAs
  • Ideal negative control for RNAi experiments using human, mouse or rat SMARTpool reagents.