Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon ON-TARGETplus™ GAPD Control siRNA
Designed and modified to minimize off-target effects and improve silencing specificity
Manufacturer: ge healthcare dharmacon  D-001830-01-05
 View more versions of this product
Catalog No. D0018300105
  • / Each
Description & Specifications
Specifications
For Use With (Application) Validated positive controls siRNA for guaranteed silencing of GAPD, reduced off-targets due to the ON-TARGETplus™ dual-strand modifications, determination of optimal RNAi conditions
Quantity 5nm
Species Human
  • Validated positive controls siRNA for guaranteed silencing of GAPD (glyceraldehyde-3-phosphate dehydrogenase, also known as GAPDH) in human and mouse cells
  • Targets accession number NM_002046 (human) or NM_008084 (mouse) or XM_575242 (rat)
GAPD is an important enzyme in carbohydrate metabolism that is well conserved across the animal kingdom. This gene is abundantly expressed in most cells and because it is non-essential, knockdown of the corresponding mRNA does not affect cell viability.