Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon ON-TARGETplus™ GAPD Control siRNA
Designed and modified to minimize off-target effects and improve silencing specificity
$181.47 - $181.47
Specifications
For Use With (Application) Validated positive controls siRNA for guaranteed silencing of GAPD, reduced off-targets due to the ON-TARGETplus™ dual-strand modifications, determination of optimal RNAi conditions
View More Specs
Products
Catalog Number Mfr. No. Species Quantity Price Quantity    
D0018300105
ge healthcare dharmacon
D-001830-01-05
Human 5nm Each for $181.47
Description & Specifications
Specifications
For Use With (Application) Validated positive controls siRNA for guaranteed silencing of GAPD, reduced off-targets due to the ON-TARGETplus™ dual-strand modifications, determination of optimal RNAi conditions
  • Validated positive controls siRNA for guaranteed silencing of GAPD (glyceraldehyde-3-phosphate dehydrogenase, also known as GAPDH) in human and mouse cells
  • Targets accession number NM_002046 (human) or NM_008084 (mouse) or XM_575242 (rat)
GAPD is an important enzyme in carbohydrate metabolism that is well conserved across the animal kingdom. This gene is abundantly expressed in most cells and because it is non-essential, knockdown of the corresponding mRNA does not affect cell viability.