Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon ON-TARGETplus™ GAPD Control Pool
Designed and modified to minimize off-target effects and improve silencing specificity
$254.36 - $595.00
Specifications
For Use With (Application) Ideal positive controls for RNAi experiments using Human, Mouse or Rat SMARTpool™ reagents, pool of four highly functional, validated siRNAs for guaranteed silencing of GAPD in Human, Mouse or Rat cells
View More Specs
Products
Catalog Number Mfr. No. Species Quantity Price Quantity    
D0018301020
ge healthcare dharmacon
D-001830-10-20
Human 20nm Each for $595.00
D0018302005
ge healthcare dharmacon
D-001830-20-05
Mouse 5nm Each for $254.36
D0018301005
ge healthcare dharmacon
D-001830-10-05
Human 5nm Each for $254.36
Description & Specifications
Specifications
For Use With (Application) Ideal positive controls for RNAi experiments using Human, Mouse or Rat SMARTpool™ reagents, pool of four highly functional, validated siRNAs for guaranteed silencing of GAPD in Human, Mouse or Rat cells
  • Validated positive controls siRNA for guaranteed silencing of GAPD (glyceraldehyde-3-phosphate dehydrogenase, also known as GAPDH) in human and mouse cells
  • Targets accession number NM_002046 (human) or NM_008084 (mouse) or XM_575242 (rat)
GAPD is an important enzyme in carbohydrate metabolism that is well conserved across the animal kingdom. This gene is abundantly expressed in most cells and because it is non-essential, knockdown of the corresponding mRNA does not affect cell viability.