Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon ON-TARGETplus™ GAPD Control Pool
Designed and modified to minimize off-target effects and improve silencing specificity
Manufacturer: ge healthcare dharmacon  D-001830-20-05
 View more versions of this product
Catalog No. D0018302005
  • / Each
Description & Specifications
Specifications
For Use With (Application) Ideal positive controls for RNAi experiments using Human, Mouse or Rat SMARTpool™ reagents, pool of four highly functional, validated siRNAs for guaranteed silencing of GAPD in Human, Mouse or Rat cells
Quantity 5nm
Species Mouse
  • Validated positive controls siRNA for guaranteed silencing of GAPD (glyceraldehyde-3-phosphate dehydrogenase, also known as GAPDH) in human and mouse cells
  • Targets accession number NM_002046 (human) or NM_008084 (mouse) or XM_575242 (rat)
GAPD is an important enzyme in carbohydrate metabolism that is well conserved across the animal kingdom. This gene is abundantly expressed in most cells and because it is non-essential, knockdown of the corresponding mRNA does not affect cell viability.