Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon ON-TARGETplus™ Cyclophilin B Control Pool
Designed and modified to minimize off-target effects and improve silencing specificity
$254.36 - $595.00
Specifications
For Use With (Application) Ideal positive controls for RNAi experiments using Human, Mouse or Rat SMARTpool™ reagents, pool of four highly functional, validated siRNAs for guaranteed silencing of GAPD in Human, Mouse or Rat cells
View More Specs
Products
Catalog Number Mfr. No. Species Quantity Price Quantity    
D0018201005
ge healthcare dharmacon
D-001820-10-05
Human 5nm Each for $254.36
D0018202020
ge healthcare dharmacon
D-001820-20-20
Mouse 20nm Each for $595.00
D0018203005
ge healthcare dharmacon
D-001820-30-05
Rat 5nm Each for $254.36
Description & Specifications
Specifications
For Use With (Application) Ideal positive controls for RNAi experiments using Human, Mouse or Rat SMARTpool™ reagents, pool of four highly functional, validated siRNAs for guaranteed silencing of GAPD in Human, Mouse or Rat cells
This gene is abundantly expressed in most cells and because it is non-essential, knockdown of the corresponding mRNA does not affect cell viability.
  • Ideal positive controls for RNAi experiments using human, mouse or rat SMARTpool reagents.
  • Pool of four highly functional, validated siRNAs for guaranteed silencing of Cyclophilin B in human, mouse or rat cells.
  • Target accession numbers NM_000942 (human, D-001820-10), NM_011149 (mouse, D-001820-20) or NM_022536 (rat, D-001820-30). Also known as peptidylprolyl isomerase B, cyclophilin B is associated with the secretory pathway.