Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon ON-TARGETplus™ Cyclophilin B Control Pool
Designed and modified to minimize off-target effects and improve silencing specificity
Manufacturer: ge healthcare dharmacon  D-001820-30-05
 View more versions of this product
Catalog No. D0018203005
  • / Each
Description & Specifications
Specifications
For Use With (Application) Ideal positive controls for RNAi experiments using Human, Mouse or Rat SMARTpool™ reagents, pool of four highly functional, validated siRNAs for guaranteed silencing of GAPD in Human, Mouse or Rat cells
Quantity 5nm
Species Rat
This gene is abundantly expressed in most cells and because it is non-essential, knockdown of the corresponding mRNA does not affect cell viability.
  • Ideal positive controls for RNAi experiments using human, mouse or rat SMARTpool reagents.
  • Pool of four highly functional, validated siRNAs for guaranteed silencing of Cyclophilin B in human, mouse or rat cells.
  • Target accession numbers NM_000942 (human, D-001820-10), NM_011149 (mouse, D-001820-20) or NM_022536 (rat, D-001820-30). Also known as peptidylprolyl isomerase B, cyclophilin B is associated with the secretory pathway.