Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon miRIDIAN™ microRNA Mimic Negative Controls
Manufacturer: ge healthcare dharmacon  CN-001000-01-05
 View more versions of this product
Catalog No. CN10000105
  • / Each
Description & Specifications
Specifications
For Use With (Application) Non-targeting inhibitors for use as controls in miRNA inhibition experiments, distinguish between specific inhibitor activity and background effects
Quantity 5nm
We offer two universal negative controls for both inhibitors and mimics that are based on the sequences of two miRNAs in C. elegans.
These miRNAs have been confirmed to have minimal sequence identity with miRNAs in human, mouse and rat. We recommend using the mature sequence information to determine which control would be most appropriate for the miRNA being studied.
  • Negative control sequence based on cel-miR-67, mature sequence: UCACAACCUCCUAGAAAGAGUAGA and Accession Number: MIMAT0000039
  • Cel-miR-67 has been confirmed to have minimal sequence identity with miRNAs in human, mouse and rat