Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon DharmaFECT™-4 siRNA Transfection Reagent
Most broadly applicable DharmaFECT formulation for optimal siRNA or microRNA transfection into a wide range of cell types for successful RNAi experiments
Manufacturer: ge healthcare dharmacon  T-2004-03
 View more versions of this product
Catalog No. T200403
  • / Each
Description & Specifications
Specifications
Concentration 1mg/mL
For Use With (Application) High efficiency of siRNA and microRNA delivery with minimal impact on cell viability across a broad range of cell types for successful RNAi experiments
Product Type DharmaFECT-4 Reagent
Quantity 1.5mL
  • Optimized for small RNA (siRNA or microRNA) delivery
  • DharmaFECT provides four specialized lipid-based formulations for successful delivery to more cell lines than any other single reagent
  • DharmaFECT 1 is the most all-purpose transfection reagent, demonstrating efficient, low-toxicity delivery to over 80% of validated cell types
  • DharmaFECT 2, 3, and 4 offer distinct formulations to support a wider range of cell types, and permit more thorough optimization of transfection for high-value experiments and screening projects