Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon DharmaFECT™ Duo Transfection Reagents
Reagents optimized for delivery of both siRNA and plasmid in a single formulation
$186.56 - $897.44
Specifications
For Use With (Application) Delivery of both siRNA and plasmid in a single formulation for RNAi screening and reporter systems
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    
T201001
ge healthcare dharmacon
T-2010-01
0.2mL Each for $186.56
T201002
ge healthcare dharmacon
T-2010-02
0.75mL Each for $539.49
T201003
ge healthcare dharmacon
T201003
1.5mL Each for $897.44
Description & Specifications
Specifications
For Use With (Application) Delivery of both siRNA and plasmid in a single formulation for RNAi screening and reporter systems
  • Efficient delivery of siRNA (assayed by target gene knockdown) and plasmid (assayed by level of reporter expression) for RNAi screening and reporter systems
  • Good transfection results without compromising cell viability
  • Potent target knockdown together with plasmid expression