Please login to your online account to display your discounted pricing

GE Healthcare Dharmacon DharmaFECT™ Duo Transfection Reagents


Reagents optimized for delivery of both siRNA and plasmid in a single formulation

Manufacturer: ge healthcare dharmacon  T-2010-02

 View more versions of this product

Catalog No. T201002

  • / Each
 

Description & Specifications

Specifications

Concentration 1mg/mL
For Use With (Application) Delivery of both siRNA and plasmid in a single formulation for RNAi screening and reporter systems
Quantity 0.75mL

  • Efficient delivery of siRNA (assayed by target gene knockdown) and plasmid (assayed by level of reporter expression) for RNAi screening and reporter systems
  • Good transfection results without compromising cell viability
  • Potent target knockdown together with plasmid expression