Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon Accell™ siRNA Positive Control Reagents
non-species-specific controls for targeting of reporter genes or other experimental needs
Manufacturer: ge healthcare dharmacon  P-002048-01-20
 View more versions of this product
Catalog No. P0020480120
  • / Each
Description & Specifications
Specifications
For Use With (Application) Use in any mammalian cell type
Product Type siRNA Positive Control
Quantity 20nm
High-quality siRNA to target common reporter genes like β-galactosidase, luciferase, and GFP.