Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon Accell™ siRNA Positive Control Reagents
non-species-specific controls for targeting of reporter genes or other experimental needs
$476.49 - $476.49
Specifications
Product Type siRNA Positive Control
View More Specs
Products
Catalog Number Mfr. No. For Use With (Application) Quantity Price Quantity    
P0020480120
ge healthcare dharmacon
P-002048-01-20
Use in any mammalian cell type 20nm Each for $476.49
Description & Specifications
Specifications
Product Type siRNA Positive Control
High-quality siRNA to target common reporter genes like β-galactosidase, luciferase, and GFP.