Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon Accell™ siRNA Delivery Media
Specially formulated and optimized for Accell siRNA delivery
Manufacturer: ge healthcare dharmacon  B005000500
 View more versions of this product
Catalog No. B005000500
  • / Each
Description & Specifications
Specifications
For Use With (Application) Accell™ siRNA reagents which provides appropriate Serum-free conditions for Accell™ uptake while maintaining cell health
Quantity 500mL
  • Required for use with Accell siRNA reagents
  • Consistently shows strong delivery, knockdown, and high cell viability when used with Accell siRNA
  • Has performed successfully in every cell type tested to date
  • Serum-free
Use of an alternate media may degrade the performance of Accell siRNA, as it has been determined that many media formulations contain biological derivatives that interfere with its uptake and silencing.