Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon Accell™ Nontargeting Control siRNA and Nontargeting Pool
Designed to aid in the interpretation of experiments with Accell siRNA reagents as highly reliable controls for assessment of baseline cellular responses
Manufacturer: ge healthcare dharmacon  D0019100120
 View more versions of this product
Catalog No. D0019100120
  • / Each
Description & Specifications
Specifications
For Use With (Application) Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable controls for assessment of baseline cellular responses
Product Type Nontargeting siRNA No. 1 Control siRNA
Quantity 20nm
  • Designed, modified, and microarray-confirmed to have minimal targeting of known genes in human, mouse and rat cells
  • Should be included in RNAi experiments as a control for nonsequence-specific effects