Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon Accell™ Nontargeting Control siRNA and Nontargeting Pool
Designed to aid in the interpretation of experiments with Accell siRNA reagents as highly reliable controls for assessment of baseline cellular responses
$218.67 - $550.38
Specifications
For Use With (Application) Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable controls for assessment of baseline cellular responses
View More Specs
Products
Catalog Number Mfr. No. Product Type Quantity Price Quantity    
D0019100105
ge healthcare dharmacon
D0019100105
Nontargeting siRNA No. 1 Control siRNA 5nm Each for $218.67
D0019100120
ge healthcare dharmacon
D0019100120
Nontargeting siRNA No. 1 Control siRNA 20nm Each for $550.38
D0019100205
ge healthcare dharmacon
D0019100205
Nontargeting siRNA No. 2 Control siRNA 5nm Each for $218.67
D0019100305
ge healthcare dharmacon
D0019100305
Nontargeting siRNA No. 3 Control siRNA 5nm Each for $218.67
D0019100405
ge healthcare dharmacon
D0019100405
Nontargeting siRNA No. 4 Control siRNA 5nm Each for $218.67
Description & Specifications
Specifications
For Use With (Application) Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable controls for assessment of baseline cellular responses
  • Designed, modified, and microarray-confirmed to have minimal targeting of known genes in human, mouse and rat cells
  • Should be included in RNAi experiments as a control for nonsequence-specific effects