Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon Accell™ GAPD Control siRNA and GAPD Pool
Recommended as a positive silencing control
$380.81 - $380.81
Specifications
For Use With (Application) Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable positive controls for delivery and silencing capability
View More Specs
Products
Catalog Number Mfr. No. Accession Number Product Type Quantity Price Quantity    
D0019300320
ge healthcare dharmacon
D0019300320
XM_575242 GAPD Control siRNA—Rat 20nm Each for $380.81
Description & Specifications
Specifications
For Use With (Application) Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable positive controls for delivery and silencing capability
GAPD (glyceraldehyde-3-phosphate dehydrogenase, also known as GAPDH) is a well-conserved enzyme in carbohydrate metabolism. This gene is abundantly expressed in most cells and because it is nonessential, knockdown of the corresponding mRNA does not affect cell viability.
  • Designed to aid in the interpretation of experiments with Accell siRNA reagents as highly reliable positive controls for delivery and silencing capability
  • Targets accession number NM_002046 (human) or NM_008085 (mouse) or XM_575242 (rat)