Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon Accell™ GAPD Control siRNA and GAPD Pool
Recommended as a positive silencing control
Manufacturer: ge healthcare dharmacon  D0019300320
 View more versions of this product
Catalog No. D0019300320
  • / Each
Description & Specifications
Specifications
Accession Number XM_575242
For Use With (Application) Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable positive controls for delivery and silencing capability
Product Type GAPD Control siRNA—Rat
Quantity 20nm
GAPD (glyceraldehyde-3-phosphate dehydrogenase, also known as GAPDH) is a well-conserved enzyme in carbohydrate metabolism. This gene is abundantly expressed in most cells and because it is nonessential, knockdown of the corresponding mRNA does not affect cell viability.
  • Designed to aid in the interpretation of experiments with Accell siRNA reagents as highly reliable positive controls for delivery and silencing capability
  • Targets accession number NM_002046 (human) or NM_008085 (mouse) or XM_575242 (rat)